Learn More
SPS Medical 60DEG C INCUBATOR W/13MM WELL
Catalog No. NC9001913
Encompass_Preferred
Dry block incubator for use with biological indicators and calibrated for 60 deg.C for steam and chemical vapor. 13mm wells.
Catalog No. NC9001913 Supplier SPS Medical Supplier No. NDB056
Dry block incubator for use with biological indicators and calibrated for 60 deg.C for steam and chemical vapor. 13mm wells.