Learn More
MSC Eagle Flammable Safety Cabinet, 60 gal., Manual Close, 1 ea
Catalog No. 502886286
Encompass
This 60 gal. flammable liquid safety cabinet is constructed of 18-gauge steel, featuring double-walled construction and manual closing doors. It is designed for protected storage of flammable and combustible liquids, meeting key safety and compliance standards.
  • Reinforced double wall construction with 1-1/2 in air space
  • High gloss powder coat finish with red trilingual warning
  • Grounding attachment
  • Three-point latch system with key lock
  • Two-in raised, leakproof door sill
  • Adjustable shelves on 3 in centers, each supporting 350 lbs
  • Two 2 in internal vents with fire baffle and cap
  • Leveling legs
  • Complies with NFPA Code 30, OSHA, and is FM Approved
Catalog No. 50-288-6286 Supplier MSC Supplier No. 16816407
This 60 gal. flammable liquid safety cabinet is constructed of 18-gauge steel, featuring double-walled construction and manual closing doors. It is designed for protected storage of flammable and combustible liquids, meeting key safety and compliance standards.
  • Reinforced double wall construction with 1-1/2 in air space
  • High gloss powder coat finish with red trilingual warning
  • Grounding attachment
  • Three-point latch system with key lock
  • Two-in raised, leakproof door sill
  • Adjustable shelves on 3 in centers, each supporting 350 lbs
  • Two 2 in internal vents with fire baffle and cap
  • Leveling legs
  • Complies with NFPA Code 30, OSHA, and is FM Approved