Learn More
Foxx Life Sciences 60L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. 
GREENER_CHOICE
Catalog No. NC2213494
Encompass_Preferred
60L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 10L, 13.5L, and 20L. The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.
This item is not returnable. View return policy
Catalog No. NC2213494 Supplier Foxx Life Sciences Supplier No. 3922011OEM
60L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 10L, 13.5L, and 20L. The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.