Learn More
Chemglass Life Sciences 60mL Addition Funnel, Graduated, 19/22 Joint Size, 2mm PTFE Metering Stpk, 230mm OAH
SDP
Catalog No. CG171311
Encompass_Preferred
60mL Addition Funnel, Graduated, 19/22 Joint Size, 2mm PTFE Metering Stopcock, 230mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171311 Supplier Chemglass Life Sciences Supplier No. CG171311
60mL Addition Funnel, Graduated, 19/22 Joint Size, 2mm PTFE Metering Stopcock, 230mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.