Learn More
Chemglass Life Sciences 60mL Addition Funnel, 14/20 Joint Size, 2mm Stpk, 0-2mm PTFE Valve, 235mm OAH
SDP
Catalog No. CG170412
Encompass_Preferred
60mL Addition Funnel, 14/20 Joint Size, 2mm Stopcock, 0-2mm PTFE Valve, 235mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG170412 Supplier Chemglass Life Sciences Supplier No. CG170412
60mL Addition Funnel, 14/20 Joint Size, 2mm Stopcock, 0-2mm PTFE Valve, 235mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.