Learn More
Chemglass Life Sciences 60mL Addition Funnel, 24/40 Joint Size, 2mm Glass Stpk, 265mm OAH
SDP
Catalog No. 50873259
Encompass_Preferred
60mL Addition Funnel, 24/40 Joint Size, 2mm Glass Stopcock, 265mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. For replacement stoppers see CG-3000.
Catalog No. 50-873-259 Supplier Chemglass Life Sciences Supplier No. CG170001
60mL Addition Funnel, 24/40 Joint Size, 2mm Glass Stopcock, 265mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. For replacement stoppers see CG-3000.