Learn More
Chemglass Life Sciences 60mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm Glass Stpk, 225mm OAH
SDP
Catalog No. 502733580
Encompass_Preferred
Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 joints are supplied complete with a stopper.
Catalog No. 50-273-3580 Supplier Chemglass Life Sciences Supplier No. CG171019
Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 joints are supplied complete with a stopper.