Learn More
C&G Containers 60 mL Amber Boston Round Glass Bottles, Excellent UV Protection, Wide Chemical Compatibility, 100/Case
SDP
Catalog No. NC1922668
Encompass_Preferred
The 60 mL Amber Boston Round Glass Bottles with White Polypropylene Caps are designed for effective storage and protection of light-sensitive samples, chemicals, and solvents. These bottles offer excellent UV protection, a narrow screw-neck opening, and are ideal for general laboratory use, ensuring cleanliness with convenient packing. Their robust material and design set them apart for safer storage solutions in various applications.
Catalog No. NC1922668 Supplier C&G Containers Supplier No. LY93A020A01BZS1
The 60 mL Amber Boston Round Glass Bottles with White Polypropylene Caps are designed for effective storage and protection of light-sensitive samples, chemicals, and solvents. These bottles offer excellent UV protection, a narrow screw-neck opening, and are ideal for general laboratory use, ensuring cleanliness with convenient packing. Their robust material and design set them apart for safer storage solutions in various applications.