Learn More
Environmental Sampling Supply Inc 60ML AMBER CLOSED TOP VL 72/CS
Catalog No. NC1173519
Encompass_Preferred
60ML AMBER BOROSILICATE GLASS VIAL/24MMPOLY CLOSEDTOP CLOSURE/TEFLON LINER, EPA CERTIFIED
Catalog No. NC1173519 Supplier Environmental Sampling Supply Inc Supplier No. 72600410QC
60ML AMBER BOROSILICATE GLASS VIAL/24MMPOLY CLOSEDTOP CLOSURE/TEFLON LINER, EPA CERTIFIED