Learn More
LUKNOVA INC Easyfil Filter 60Ml, 24/25 Joint Adaptor, Empty, Conical Shape, Polyethylene Frit Pore Size 10µm. Integrated Via Welding As One-Piece Leak-Free Filter. Polypropylene Structure Resistant To Common Solvents. Temperature Upto 120C. 100 Pieces Per Pack.
Catalog No. NC0627126
Encompass
Luknova Patented Easyfil Filter. 60Ml Volume, 24/25 Joint Adaptor To Fit Standard Flask And Side-Arm To Connect Vacumm, Empty, Conical Shape, Polyethylene Frit Pore Size 10µm. This Unit Comes At 100 Filters Per Pack. Frit, Adaptor, And Funnel Body Were Integrated Via Welded Seals As One-Piece Leak-Free Filter. Polypropylene Structure Resistant To Common Aqueous And Organic Solvents Like Ethyl Acetate , Dimethylformamide (Dmf), Dimethylamine (Dma), Diethyl Ether (Dme), Dichloromethane (Dcm), Tetrahydrofuran (Thf), Hexane, Concentrated Sulfuric Acid (H2So4), Hydrochloric Acid (Hcl), Hydrobromic Acid (Hbr), Hydrogen Iodide (Hi), Acetic Acid, Trifluoroacetic Acid (Tfa), Saturated Naoh And Koh Solution, And Even Dimethyl Sulfoxide (Dmso) At Temperature ≤120C (248F). Safe And Ready To Use For Faster Filtration Without Contamination, Save Time And Solvent, Low-Cost Disposable And Resuable Up To 20 Times. Welded Seals Eliminating Bypass Leakage And Long-Stem Minimizing Foaming, Splashing.
Catalog No. NC0627126 Supplier LUKNOVA INC Supplier No. SX820021
Luknova Patented Easyfil Filter. 60Ml Volume, 24/25 Joint Adaptor To Fit Standard Flask And Side-Arm To Connect Vacumm, Empty, Conical Shape, Polyethylene Frit Pore Size 10µm. This Unit Comes At 100 Filters Per Pack. Frit, Adaptor, And Funnel Body Were Integrated Via Welded Seals As One-Piece Leak-Free Filter. Polypropylene Structure Resistant To Common Aqueous And Organic Solvents Like Ethyl Acetate , Dimethylformamide (Dmf), Dimethylamine (Dma), Diethyl Ether (Dme), Dichloromethane (Dcm), Tetrahydrofuran (Thf), Hexane, Concentrated Sulfuric Acid (H2So4), Hydrochloric Acid (Hcl), Hydrobromic Acid (Hbr), Hydrogen Iodide (Hi), Acetic Acid, Trifluoroacetic Acid (Tfa), Saturated Naoh And Koh Solution, And Even Dimethyl Sulfoxide (Dmso) At Temperature ≤120C (248F). Safe And Ready To Use For Faster Filtration Without Contamination, Save Time And Solvent, Low-Cost Disposable And Resuable Up To 20 Times. Welded Seals Eliminating Bypass Leakage And Long-Stem Minimizing Foaming, Splashing.