Learn More
Chemrus Inc 60ML FILTER FUNNEL 1.5G CELITE
Supplier:  Chemrus Inc CR103840
Encompass
Chemrus 60 mL polypropylene barrel-shaped disposable filter funnel, packed with 1.5 gram Celite
Catalog No. 50-823-15