Learn More
Chemglass Life Sciences 60mL Filter Funnel, 14/20 Joint Size, 30mm OD Coarse Frit, Valve On Side Arm, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF054320
Encompass_Preferred
60mL Filter Funnel, 14/20 Joint Size, 30mm OD Coarse Frit, Valve On Side Arm, Airfree, Schlenk. Similar to AF-0542 but with the top having a standard taper inner joint instead of the outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. AF054320