Learn More
C&G Containers 60ML FLT GLS SS WHT PTFE A1BCL
SDP
Catalog No. NC9842925
Encompass_Preferred
Jar, Glass; Straight Sided; 2 oz.; Flint, Neck Size: 53 to 400, 24/CS
This item is not returnable. View return policy
Catalog No. NC9842925 Supplier C&G Containers Supplier No. LFS0022000
Jar, Glass; Straight Sided; 2 oz.; Flint, Neck Size: 53 to 400, 24/CS