Learn More
Thermco Products Inc 60ml PETG plastic bottle w/Biosafe Liquid & Magnets
Catalog No. NC0600312
Encompass_Preferred
60ml bottle with insulating liquid for liquid-in-glass thermometer and/or temperature probes to monitor temperatures in freezers, refrigerators, incubators, oven and other laboratory instruments. Bottle is filled with bio-safe glycol and has dual magnets to install on doors or wall of the instrument saving space
Catalog No. NC0600312 Supplier Thermco Products Inc Supplier No. ACCBOT60BIO
60ml bottle with insulating liquid for liquid-in-glass thermometer and/or temperature probes to monitor temperatures in freezers, refrigerators, incubators, oven and other laboratory instruments. Bottle is filled with bio-safe glycol and has dual magnets to install on doors or wall of the instrument saving space