Learn More
Ace Glass, Inc. 60mL separatory funnel with locking stopper
SDP
Catalog No. 50974698
Encompass_Preferred
60ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-698 Supplier Ace Glass, Inc. Supplier No. 723155
60ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.