Learn More
Foxx Life Sciences 60mL Wide Mouth PP Reagent Bottles, 72CS
GREENER_CHOICE
Catalog No. 5000846671
Encompass_Preferred
EZLabpure™  60mL PP Reagent Bottles Autoclavable, Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Wide Mouth PP Reagent Bottle features large openings for easy filling, cleaning & rapid content transfer. PP Chemical Reagent Bottles are rigid and fully autoclavable without deforming making them durable and reliable for storing aqueous laboratory solutions.
Features
Autoclavable up to 121°C
PP Storage Reagent Bottles are rigid and translucent for easy content visibility.
PP Storage Bottles offer excellent resistance to aqueous solutions and non-corrosive chemicals.
Leak-proof Linerless screw caps for secure sealing.
PP Bottles with Low Liquid Retention
Catalog No. 50-008-46671 Supplier Foxx Life Sciences Supplier No. 7652811FLS
EZLabpure™  60mL PP Reagent Bottles Autoclavable, Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Wide Mouth PP Reagent Bottle features large openings for easy filling, cleaning & rapid content transfer. PP Chemical Reagent Bottles are rigid and fully autoclavable without deforming making them durable and reliable for storing aqueous laboratory solutions.
Features
Autoclavable up to 121°C
PP Storage Reagent Bottles are rigid and translucent for easy content visibility.
PP Storage Bottles offer excellent resistance to aqueous solutions and non-corrosive chemicals.
Leak-proof Linerless screw caps for secure sealing.
PP Bottles with Low Liquid Retention