Learn More
CELLTREAT Scientific Products 60mL Wide Mouth Bottle, Round, PP, Non-sterile, 48/CS
SDP
Catalog No. 501535080
Encompass_Preferred
CELLTREAT Scientific Products 60mL Wide Mouth Bottle. Round shape and easy-pour wide mouth design. Bottle and cap manufactured from virgin polypropylene for excellent chemical resistance with durable, rigid walls. Features liner-less leak proof screw top closure. Bottle and cap come fully assembled. Ideal for specimen and chemical storage, storage of laboratory reagents and biological buffers and for sample shipment and packaging. Non-sterile; autoclavable. Every case is printed with the lot number for quality traceability. Individually packed in bags, 24/case.
Catalog No. 50-153-5080 Supplier CELLTREAT Scientific Products Supplier No. 229794
CELLTREAT Scientific Products 60mL Wide Mouth Bottle. Round shape and easy-pour wide mouth design. Bottle and cap manufactured from virgin polypropylene for excellent chemical resistance with durable, rigid walls. Features liner-less leak proof screw top closure. Bottle and cap come fully assembled. Ideal for specimen and chemical storage, storage of laboratory reagents and biological buffers and for sample shipment and packaging. Non-sterile; autoclavable. Every case is printed with the lot number for quality traceability. Individually packed in bags, 24/case.