Learn More
Chemglass Life Sciences Reagent Reservoir, 60mL, with Lid/Cover, Non-Sterile, 5/Pk, 25/Case
SDP
Catalog No. 501897721
Encompass_Preferred
V-shape Bottom, used for cell culture and immunoassay experimentsFor single and multi-channel pipettesUSP VI PP materialReusable, autoclave at 121℃Special designed lid to prevent contaminationClear graduationsDNase/RNase free, Non-pyrogenic
Catalog No. 50-189-7721 Supplier Chemglass Life Sciences Supplier No. CGN3796005
V-shape Bottom, used for cell culture and immunoassay experimentsFor single and multi-channel pipettesUSP VI PP materialReusable, autoclave at 121℃Special designed lid to prevent contaminationClear graduationsDNase/RNase free, Non-pyrogenic