Learn More
Thermco Products Inc 60ml PETG plastic bottle w/sand
Catalog No. NC1270750
Encompass_Preferred
60ml bottle with insulating sand for liquid-in-glass thermometer and/or temperature probes to monitor temperatures in low temp freezer & oven and other laboratory instruments. Bottle is filled with sand and has dual magnets to install on doors or wall of the instrument saving space
Catalog No. NC1270750 Supplier Thermco Products Inc Supplier No. ACCBOT60SD
60ml bottle with insulating sand for liquid-in-glass thermometer and/or temperature probes to monitor temperatures in low temp freezer & oven and other laboratory instruments. Bottle is filled with sand and has dual magnets to install on doors or wall of the instrument saving space