Learn More
C&G Containers 60 mL Clear Borosilicate Glass Vial, Precleaned, Polypropylene Closure
SDP
Catalog No. NC3735429
Encompass_Preferred
60 mL Clear Borosilicate Glass Vial: This high-purity vial is designed for sensitive analytical applications, ensuring minimal TOC contribution (<20 ppb) during use with 85% phosphoric acid. Constructed from Type I borosilicate glass, it offers superior chemical resistance and sample integrity. It includes a polypropylene closure and bonded PTFE/silicon septum, making it ideal for reliable sample collection and storage.
Catalog No. NC3735429 Supplier C&G Containers Supplier No. LFV060786300
60 mL Clear Borosilicate Glass Vial: This high-purity vial is designed for sensitive analytical applications, ensuring minimal TOC contribution (<20 ppb) during use with 85% phosphoric acid. Constructed from Type I borosilicate glass, it offers superior chemical resistance and sample integrity. It includes a polypropylene closure and bonded PTFE/silicon septum, making it ideal for reliable sample collection and storage.