Learn More
Ace Glass, Inc. 60ML VOLUMETRIC BULB 60ML VOLUMETRIC BULB
SDP
Catalog No. 50975877
Encompass_Preferred
60mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-975-877 Supplier Ace Glass, Inc. Supplier No. 800444
60mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.