Learn More
Chemglass Life Sciences 60mm Cell Culture Dish, TC, Sterile
SDP
Catalog No. 501897566
Encompass
60mm Cell Culture Dish, TC, Sterile. Cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.
Catalog No. 50-189-7566 Supplier Chemglass Life Sciences Supplier No. CGE1800060
60mm Cell Culture Dish, TC, Sterile. Cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.