Learn More
Chemglass Life Sciences 60mm Cell Culture Dish, TC, Sterile, 20/bag, 500/cs
SDP
Catalog No. 501897642
Encompass_Preferred
NEST cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.Made from high clarity, 100% virgin polystyreneStackable for easy storage and handlingFlat transparent surface for distortion-free observationSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityNon-Pyrogenic, DNase/RNase Free
Catalog No. 50-189-7642 Supplier Chemglass Life Sciences Supplier No. CGN1805060
NEST cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.Made from high clarity, 100% virgin polystyreneStackable for easy storage and handlingFlat transparent surface for distortion-free observationSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityNon-Pyrogenic, DNase/RNase Free