Learn More
Tritech Research Inc 60MM NONVENTED P.DISHES 500/CS
SDP
Catalog No. NC9321999
Encompass
60Mm Non-Vented, Sharp Edge Petri Dishes (500/Cs). 60Mm Sterile Polystyrene Petri Dishes, Non-Vented To Prevent Airflow. Stackable, Sturdy, Super-Flat, And Crystal Clear. Sharp Corner On Bottom Edge. Packed 500 Per Case In Sterile Sleeves. These Petri Dishes Have Lids Are Not Raised. By Reducing Ventilation To The Dish, Samples Take Much Longer To Dry Out And The Chances Of External Contamination Are Further Reduced. Compared To Our Model T3306, Which Are Also "60Mm Non-Vented Petri Dishes", These Are Slightly Smaller And Lower Profile. These Are The Top Choice Of C. Elegans Labs.
Catalog No. NC9321999 Supplier Tritech Research Inc Supplier No. T3308P
60Mm Non-Vented, Sharp Edge Petri Dishes (500/Cs). 60Mm Sterile Polystyrene Petri Dishes, Non-Vented To Prevent Airflow. Stackable, Sturdy, Super-Flat, And Crystal Clear. Sharp Corner On Bottom Edge. Packed 500 Per Case In Sterile Sleeves. These Petri Dishes Have Lids Are Not Raised. By Reducing Ventilation To The Dish, Samples Take Much Longer To Dry Out And The Chances Of External Contamination Are Further Reduced. Compared To Our Model T3306, Which Are Also "60Mm Non-Vented Petri Dishes", These Are Slightly Smaller And Lower Profile. These Are The Top Choice Of C. Elegans Labs.