Learn More
Andwin Scientific 60mm TPP Dishes, 840/cs
SDP
Catalog No. NC1665152
Encompass
Premium TPP Tissue Culture Dishes. 60 mm TC Dish (22.1 cm² growth surface).
Catalog No. NC1665152 Supplier Andwin Scientific Supplier No. TP93060
Premium TPP Tissue Culture Dishes. 60 mm TC Dish (22.1 cm² growth surface).