Learn More
Thomas Scientific 60mm x 15mm Non-Treated Petri Dishes w/Grip Ring, Sterile
SDP
Catalog No. 501461526
Encompass
CELLTREAT Tissue Culture Dishes are manufactured under the most stringent conditions in order to assure the integrity of your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used for CELLTREAT products produces a negatively charged, hydrophilic surface for consistent cell attachment. Growth surfaces are flat and free from striation to maximize usable growth area and transparency. Dishes are available in four different sizes; 35mm, 70mm, 90mm and 150mm. Uniformly molded plastic bottom provides a distortion free bottom surface for consistent cell attachment. All dishes are supplied with lids. Lids contain molded-in spacers to provide consistent gas exchange and stacking rings that add stability for handling tall stacks of dishes. The 35mm, 70mm and 90mm dishes are packaged in easy o
Catalog No. 50-146-1526 Supplier Thomas Scientific Supplier No. 1213F04
CELLTREAT Tissue Culture Dishes are manufactured under the most stringent conditions in order to assure the integrity of your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used for CELLTREAT products produces a negatively charged, hydrophilic surface for consistent cell attachment. Growth surfaces are flat and free from striation to maximize usable growth area and transparency. Dishes are available in four different sizes; 35mm, 70mm, 90mm and 150mm. Uniformly molded plastic bottom provides a distortion free bottom surface for consistent cell attachment. All dishes are supplied with lids. Lids contain molded-in spacers to provide consistent gas exchange and stacking rings that add stability for handling tall stacks of dishes. The 35mm, 70mm and 90mm dishes are packaged in easy o