Learn More
Bellco Glass, Inc. 60mm x 15mm Petri Dish
Supplier: Bellco Glass, Inc. 229663000
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions. Petri Nontreated Cell Culture Dishes have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications. 60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid With griping feature Working Volume 78mL Working Area 21cm2 Case Quantity 500