Learn More
CELLTREAT Scientific Products 60mm x 15mm Petri Dish, Sterile
SDP
Catalog No. 50828744
Encompass_Preferred
CELLTREAT Scientific Products 60mm x 15mm Non-Treated Petri Dish. CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Lids contain stacking rings for increased stability. Working volume of 7-8mL and growth area of 21cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50-828-744 Supplier CELLTREAT Scientific Products Supplier No. 229665
CELLTREAT Scientific Products 60mm x 15mm Non-Treated Petri Dish. CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Lids contain stacking rings for increased stability. Working volume of 7-8mL and growth area of 21cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.