Learn More
Medicus Health 60 Place, Stainless Steel Staining Rack with Handle
SDP
Catalog No. NC3358412
Encompass
60 Place Stainlesss Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned
∙ Holds slides size 3"L x 1"W, 1.5"W, 2"W
∙ 60 Place Stainlesss Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides
∙ Great for Hematology Departments who need to do Wright's Stains
∙ Slide Staining Dipper is sold by the each
∙ Slide Dipper measures: 7.6″W x 3.5″D x 3″H
∙ Material: Stainless steel
Catalog No. NC3358412 Supplier Medicus Health Supplier No. 5441M5
60 Place Stainlesss Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned
∙ Holds slides size 3"L x 1"W, 1.5"W, 2"W
∙ 60 Place Stainlesss Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides
∙ Great for Hematology Departments who need to do Wright's Stains
∙ Slide Staining Dipper is sold by the each
∙ Slide Dipper measures: 7.6″W x 3.5″D x 3″H
∙ Material: Stainless steel