Learn More
Air Science Usa Forensic Evidence Drying Cabinet, 60" Wide, Ductless Filtration, Tamper-Resistant, 485 lbs
SDP
Catalog No. NC3371781
Encompass_Preferred
The Safe Keeper Forensic Evidence Drying Cabinet is designed to securely dry and store forensic evidence while maintaining chain of custody integrity. With a spacious 60 in wide duplex chamber, advanced ductless filtration for odor and fume elimination, and robust compliance with Osha and ISO standards, this cabinet ensures safe handling and processing of delicate evidence. Its user-friendly features and efficient airflow system set it apart in forensic environments
Catalog No. NC3371781 Supplier Air Science Usa Supplier No. FDC008DA
Add to Cart
Add to Cart
Request a Quote
The Safe Keeper Forensic Evidence Drying Cabinet is designed to securely dry and store forensic evidence while maintaining chain of custody integrity. With a spacious 60 in wide duplex chamber, advanced ductless filtration for odor and fume elimination, and robust compliance with Osha and ISO standards, this cabinet ensures safe handling and processing of delicate evidence. Its user-friendly features and efficient airflow system set it apart in forensic environments