Learn More
Molecular Devices Molecular Devices Confocal Disk Module, Deep Tissue Geometry, Spinning Disk
Catalog No. 5000014714
Encompass_Preferred
The Molecular Devices Deep Tissue Confocal Disk is a spinning-disk confocal module engineered to improve imaging quality from thick, three-dimensional biological samples. Its specialized disk geometry suppresses out-of-focus light and reduces haze, enabling deeper penetration and clearer volumetric imaging in high-content workflows.
  • Improves suppression of out-of-focus light for clearer images.
  • Reduces haze and pinhole crosstalk in thick samples.
  • Enables deeper penetration into 3D tissue and organoid samples.
  • Optimized disk geometry for high-content confocal imaging workflows.
  • Designed for integration with automated high-throughput imaging systems.
Catalog No. 50-000-14714 Supplier Molecular Devices Supplier No. 5315513
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices Deep Tissue Confocal Disk is a spinning-disk confocal module engineered to improve imaging quality from thick, three-dimensional biological samples. Its specialized disk geometry suppresses out-of-focus light and reduces haze, enabling deeper penetration and clearer volumetric imaging in high-content workflows.
  • Improves suppression of out-of-focus light for clearer images.
  • Reduces haze and pinhole crosstalk in thick samples.
  • Enables deeper penetration into 3D tissue and organoid samples.
  • Optimized disk geometry for high-content confocal imaging workflows.
  • Designed for integration with automated high-throughput imaging systems.