Learn More
CELLTREAT Scientific Products 60mm x 15mm Tissue Culture Treated Dish, Sterile
SDP
Catalog No. 501149055
Encompass_Preferred
CELLTREAT Scientific Products 60mm x 15mm Tissue Culture Treated Dish. Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 7-8mL and growth area of 21cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/re-sealable bag, 500/case.
Catalog No. 50-114-9055 Supplier CELLTREAT Scientific Products Supplier No. 229661
CELLTREAT Scientific Products 60mm x 15mm Tissue Culture Treated Dish. Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 7-8mL and growth area of 21cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/re-sealable bag, 500/case.