Learn More
Waters Corp Centrifuge Tube Rack, 60-Tube Capacity, 15 mL, 1x2 Format
Catalog No. NC3597547 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 60 x 15 mL Centrifuge Tube Rack Domino is engineered to accommodate a large format rack, capable of securely holding up to 60 tubes. This specific Domino features a 1x2 format, meaning it occupies two horizontal positions on the Andrew+ robot workbench.
  • Dominos are machine-readable consumable holders that create the interface between various consumable types and the Andrew+ pipetting robot.
  • Enables seamless automation of numerous laboratory workflows.
  • Equipped with QR codes housing critical labware information and ARUCO codes for precise spatial positioning.
  • An indispensable component of any Andrew+ automation setup.
Catalog No. NC3597547 Supplier Waters Corp Supplier No. 186010160
The 60 x 15 mL Centrifuge Tube Rack Domino is engineered to accommodate a large format rack, capable of securely holding up to 60 tubes. This specific Domino features a 1x2 format, meaning it occupies two horizontal positions on the Andrew+ robot workbench.
  • Dominos are machine-readable consumable holders that create the interface between various consumable types and the Andrew+ pipetting robot.
  • Enables seamless automation of numerous laboratory workflows.
  • Equipped with QR codes housing critical labware information and ARUCO codes for precise spatial positioning.
  • An indispensable component of any Andrew+ automation setup.