Learn More
Ace Glass, Inc. 60mm ID X 180mm H. Cellulose extraction thimble
SDP
Catalog No. 50973430
Encompass
60mm I.D. x 180mm high Cellulose extraction thimble for giant soxhlet extraction apparatus. Cellulose fiber thimbles possess ample rigidity, good retention, and are readily permeable to the flow of ether and other organic solvents.
Catalog No. 50-973-430 Supplier Ace Glass, Inc. Supplier No. 681202
60mm I.D. x 180mm high Cellulose extraction thimble for giant soxhlet extraction apparatus. Cellulose fiber thimbles possess ample rigidity, good retention, and are readily permeable to the flow of ether and other organic solvents.