Learn More
ABclonal Technology 610 Rabbit anti-Human BCL6
SDP
Catalog No. 5000008771
Encompass_Preferred
The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal POZ domain This protein acts as a sequence-specific repressor of transcription and has been shown to modulate the transcription of STAT-dependent IL-4 responses of B cells This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors This gene is found to be frequently translocated and hypermutated in diffuse large-cell lymphoma (DLCL) and may be involved in the pathogenesis of DLCL Alternatively spliced transcript variants encoding different protein isoforms have been found for this gene
Catalog No. 50-000-08771 Supplier ABclonal Technology Supplier No. A26765200T
The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal POZ domain This protein acts as a sequence-specific repressor of transcription and has been shown to modulate the transcription of STAT-dependent IL-4 responses of B cells This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors This gene is found to be frequently translocated and hypermutated in diffuse large-cell lymphoma (DLCL) and may be involved in the pathogenesis of DLCL Alternatively spliced transcript variants encoding different protein isoforms have been found for this gene