Learn More
MSC PORTABLE DIGITAL SCALE
Catalog No. 502734558
Encompass
610 g, 0.01 g Graduation, Backlit Graphic LCD Portable Balance
Catalog No. 50-273-4558 Supplier MSC Supplier No. 65533150
610 g, 0.01 g Graduation, Backlit Graphic LCD Portable Balance