Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrohm USA 61903020 16MM STIR BAR
SDP

Catalog No. NC0313558 Shop All Metrohm USA Products
Encompass

Stirring Bar with magnetic core, PTFE coated, length 16 mm

Catalog No. NC0313558 Supplier Metrohm USA Supplier No. 61903020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stirring Bar with magnetic core, PTFE coated, length 16 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.