Learn More
Sino Biological Recombinant Sus scrofa (Pig) IL-10 Protein
Supplier:  Sino Biological 62000WNAE20
Encompass_Preferred
A DNA sequence encoding the Sus scrofa (Pig) IL10 (Q29055) (Ser19-Asn175) was expressed with an initial Met at the N-terminal.
Catalog No. 50-162-3219