Learn More
Sino Biological Recombinant Sus scrofa (Pig) IL-6 Protein
Supplier:  Sino Biological 62006WNAE20
Encompass_Preferred
A DNA sequence encoding the sus scrofa IL6 (NP_999564.1) (Pro29-Met212) was expressed with an initial Met.
Catalog No. 50-162-3229