Learn More
eMolecules​ 621-23-8 | 1,3,5-Trimethoxybenzene | Accela ChemBio (ASD) | MFCD00008385 | 168.192 | C9H12O3 | 98.000 | COc1cc(OC)cc(OC)c1 | 5g | 375960082
Catalog No. 5000159335 Shop All eMolecules Inc. Products
Encompass_Preferred
1,3,5-Trimethoxybenzene | Accela ChemBio (ASD) | 621-23-8 | MFCD00008385 | 168.192 | C9H12O3 | 98.000 | COc1cc(OC)cc(OC)c1 | 5g | 375960082
Catalog No. 50-001-59335 Supplier eMolecules​ Supplier No. 375960082
1,3,5-Trimethoxybenzene | Accela ChemBio (ASD) | 621-23-8 | MFCD00008385 | 168.192 | C9H12O3 | 98.000 | COc1cc(OC)cc(OC)c1 | 5g | 375960082