Learn More
eMolecules​ 624-08-8 | 9-Heptadecanol | Accela ChemBio (ASD) | MFCD00046734 | 256.474 | C17H36O | 97.000 | CCCCCCCCC(O)CCCCCCCC | 25g | 666597018
Catalog No. 5000158955 Shop All eMolecules Inc. Products
Encompass_Preferred
9-Heptadecanol | Accela ChemBio (ASD) | 624-08-8 | MFCD00046734 | 256.474 | C17H36O | 97.000 | CCCCCCCCC(O)CCCCCCCC | 25g | 666597018
Catalog No. 50-001-58955 Supplier eMolecules​ Supplier No. 666597018
Add to Cart
Add to Cart
9-Heptadecanol | Accela ChemBio (ASD) | 624-08-8 | MFCD00046734 | 256.474 | C17H36O | 97.000 | CCCCCCCCC(O)CCCCCCCC | 25g | 666597018