Learn More
Electronic Imaging Materials Inc 626 XyResist Polyester Label, Chemical Resistant, Gloss White, 0.875 in
SDP
Catalog No. NC3496211
Encompass
The 626 XyResist Polyester Label by Electronic Imaging Materials Inc. is a high-performance, chemical-resistant label ideal for laboratory and industrial applications. Designed from durable polyester, it offers excellent resistance to solvents and harsh environments, ensuring legibility and integrity of information. Its gloss white finish and permanent acrylic adhesive allow for reliable adhesion on various surfaces, making it perfect for asset tracking and parts identification.
Catalog No. NC3496211 Supplier Electronic Imaging Materials Inc Supplier No. 310518
The 626 XyResist Polyester Label by Electronic Imaging Materials Inc. is a high-performance, chemical-resistant label ideal for laboratory and industrial applications. Designed from durable polyester, it offers excellent resistance to solvents and harsh environments, ensuring legibility and integrity of information. Its gloss white finish and permanent acrylic adhesive allow for reliable adhesion on various surfaces, making it perfect for asset tracking and parts identification.