Learn More
MATHESON TRI GAS 6295-A EXCESS FLOW VLV NPTF
Catalog No. NC2184563
Encompass
6295-A excess flow valve NPTF 5 SLPM @ 1000 PSIG
This item is not returnable. View return policy
Catalog No. NC2184563 Supplier MATHESON TRI GAS Supplier No. SEQ 6295A
6295-A excess flow valve NPTF 5 SLPM @ 1000 PSIG