Learn More
Burch Bottle & Packaging Inc 63-400 WHITE POLYPROPYLENE P/E
Supplier:  Burch Bottle & Packaging Inc 2WP0631PX
Encompass
Stock number 2WP0631PX is a 63-400 White Polypropylene injection molded closure. This cap features ribbed sides allowing for easy application/removal and a matte top. The F217 (foam) liner inside is a general liner that is compatible with a variety of products and reduces the transmission of moisture.
Catalog No. NC1778647