Learn More
63.5/24.0/12.5 Water/Acetonitrile/Tetrahydrofuran, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| CAS | 7732-18-5, 75-05-8, 109-99-9 |
| CAS Max % | 68.00%, 20.10%, 11.90% |
| Quantity | 4 x 4 L |
| Flash Point | -3,3°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Purity Grade Notes | HPLC grade blends |
| Boiling Point | 61.1°C |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.