Learn More
Sino Biological Recombinant Danio rerio (zebrafish) EFNB2A Protein (hFc Tag)
Supplier:  Sino Biological 63002Z02H100
Encompass_Preferred
A DNA sequence encoding the zebrafish EFNB2A (O73874) (Met 1-Ala222) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3232