Learn More
Sino Biological Recombinant Danio rerio (zebrafish) BMP-2 Protein
Supplier:  Sino Biological 63003ZNAE20
Encompass_Preferred
A DNA sequence encoding the zebrafish BMP2A (B3DI86) (Gln272-Arg386) was expressed and purified with an initial Met.
Catalog No. 50-162-3237