Learn More
eMolecules​ 631915-50-9 | 3-[(Fmoc-amino)methyl]benzeneacetic acid | Acrotein ChemBio Inc. | MFCD06656460 | 387.435 | C24H21NO4 | 97.000 | OC(=O)Cc1cccc(CNC(=O)OCC2c3ccccc3-c3ccccc23)c1 | 1g | 487535540
Catalog No. 5000491355 Shop All eMolecules Inc. Products
Encompass_Preferred
3-[(Fmoc-amino)methyl]benzeneacetic acid | Acrotein ChemBio Inc. | 631915-50-9 | MFCD06656460 | 387.435 | C24H21NO4 | 97.000 | OC(=O)Cc1cccc(CNC(=O)OCC2c3ccccc3-c3ccccc23)c1 | 1g | 487535540
Catalog No. 50-004-91355 Supplier eMolecules​ Supplier No. 487535540
3-[(Fmoc-amino)methyl]benzeneacetic acid | Acrotein ChemBio Inc. | 631915-50-9 | MFCD06656460 | 387.435 | C24H21NO4 | 97.000 | OC(=O)Cc1cccc(CNC(=O)OCC2c3ccccc3-c3ccccc23)c1 | 1g | 487535540