Learn More
Biotium Phalloidin, CF633
Catalog No. NC1027461
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. C dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF633 dye has excitation/emission at 630/650 nm.
Catalog No. NC1027461 Supplier Biotium Supplier No. 00046
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. C dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF633 dye has excitation/emission at 630/650 nm.