Learn More
Refrigiwear Inc. 6345 REGULAR BLACK OSA CONVERTIBLE HEADWEAR WITH 2 FACE LAYERS
SDP
Catalog No. NC3867029
Encompass_Preferred
One of the worst winter experiences is realizing the feeling in your face and ears is long gone. In extreme temperatures, it's more important than ever to protect your head and neck from that biting cold. The 4-in-1 Convertible Balaclava provides the most customizable coverage for your face, neck, ears and head, delivering maximum flexibility as your activity level and body temperature change throughout the day. The 4-in-1 Convertible Balaclava gives you total control of the body heat from your neck, ears and head. Wear it with or without the mesh face mask liner to adjust breathability. Wear it as a gaiter when you need to cool off or when you only need extra protection around your neck. 92% Polyester/ 8% Spandex Knit brushed top layer 92%Polyester/8% Spandex mesh liner 4-in-1: wear as face mask with or without liner, as a gaiter, or as a full balaclava    
Catalog No. NC3867029 Supplier Refrigiwear Inc. Supplier No. 6345RBLKOSA
One of the worst winter experiences is realizing the feeling in your face and ears is long gone. In extreme temperatures, it's more important than ever to protect your head and neck from that biting cold. The 4-in-1 Convertible Balaclava provides the most customizable coverage for your face, neck, ears and head, delivering maximum flexibility as your activity level and body temperature change throughout the day. The 4-in-1 Convertible Balaclava gives you total control of the body heat from your neck, ears and head. Wear it with or without the mesh face mask liner to adjust breathability. Wear it as a gaiter when you need to cool off or when you only need extra protection around your neck. 92% Polyester/ 8% Spandex Knit brushed top layer 92%Polyester/8% Spandex mesh liner 4-in-1: wear as face mask with or without liner, as a gaiter, or as a full balaclava    